missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid hydrate, 98%

Product Code. 11493247
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493247 500 g 500g
11483247 100 g 100g
11403257 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493247 Supplier Thermo Scientific Alfa Aesar Supplier No. A16180.36
 Restricted Item
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolicy

CAS: 62708-56-9 | C18H14O8 | 358.30 g/mol

(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.

Solubility
Soluble in methanol, acetone and organic solvents. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 62708-56-9
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00149119
InChI Key YONLFQNRGZXBBF-OKILXGFUSA-N
Synonym 2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
PubChem CID 44119738
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Tekniske data

Melting Point 90°C to 92°C
Solubility Information Soluble in methanol,acetone and organic solvents. Slightly soluble in water.
IUPAC Name (2R,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate
Formula Weight 376.33 (358.31 Anhydrous)
Percent Purity ≥98%
Quantity 500 g
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid monohydrate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.