missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-Tartaric Acid Monohydrate, TRC

Product Code. 30375112
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375112 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375112 Supplier Toronto Research Chemicals Supplier No. TRCD4228155G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-Dibenzoyl-L-Tartaric Acid Monohydrate
CAS 62708-56-9
Molecular Formula C18H14O8 . H2O
Quantity 5 g
Synonym (-)-O,O-Dibenzoyl-L-Tartaric Acid Monohydrate
InChI Formula InChI=1S/C18H14O8.H2O/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12;/h1-10,13-14H,(H,19,20)(H,21,22);1H2/t13-,14-;/m1./s1
SMILES O.OC(=O)[C@H](OC(=O)c1ccccc1)[C@@H](OC(=O)c2ccccc2)C(=O)O
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid;hydrate
Molecular Weight (g/mol) 376.31
Formula Weight 376.0794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.