missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl azodicarboxylate, 94%

Product Code. 10464653 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10464653 5 g Glass bottle 5g
10329150 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10464653 Supplier Thermo Scientific Acros Supplier No. 270270050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2449-05-0 | C16H14N2O4 | 298.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2449-05-0
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.29
MDL Number MFCD00016737
InChI Key IRJKSAIGIYODAN-ISLYRVAYSA-N
Synonym dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
PubChem CID 5387121
IUPAC Name benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
SMILES C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 44°C to 52°C
Color Yellow to Orange
Flash Point >230°C
Loss on Drying 0.75% max.
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 5 g
Fieser 15,111
Solubility Information Solubility in water: insoluble
Formula Weight 298.29
Percent Purity 94%
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl azodicarboxylate, 94%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dibenzyl azodicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.