missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl azodicarboxylate, 96%

Product Code. 11364266
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364266 25 g 25g
11354266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364266 Supplier Thermo Scientific Alfa Aesar Supplier No. L19347.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2449-05-0 | C16H14N2O4 | 298.298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2449-05-0
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.298
MDL Number MFCD00016737
InChI Key IRJKSAIGIYODAN-ISLYRVAYSA-N
Synonym dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
PubChem CID 5387121
IUPAC Name benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
SMILES C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 43°C to 47°C
Flash Point >110°C (230°F)
Linear Formula C6H5CH2O2C-N=N-CO2CH2C6H5
Quantity 25 g
Beilstein 2298734
Sensitivity Light sensitive
Formula Weight 298.3
Percent Purity 96%
Chemical Name or Material Dibenzyl azodicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.