missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Azodicarboxylate, TRC

Product Code. 30355264
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30355264 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355264 Supplier Toronto Research Chemicals Supplier No. TRCD4172151G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibenzyl Azodicarboxylate
CAS 2449-05-0
Molecular Formula C16H14N2O4
Quantity 1 g
Synonym Diazenedicarboxylic Acid Bis(phenylmethyl) Ester, Azodi-Formic Acid Dibenzyl Ester, Dibenzyl Azodicarboxylate, NSC 620564
InChI Formula InChI=1S/C16H14N2O4/c19-15(21-11-13-7-3-1-4-8-13)17-18-16(20)22-12-14-9-5-2-6-10-14/h1-10H,11-12H2
SMILES O=C(OCc1ccccc1)N=NC(=O)OCc2ccccc2
IUPAC Name benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
Molecular Weight (g/mol) 298.29
Formula Weight 298.0954
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.