missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Disulphide, TRC

Product Code. 30368377
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368377 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368377 Supplier Toronto Research Chemicals Supplier No. TRCD41708010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Disulfide
CAS 150-60-7
Molecular Formula C14 H14 S2
Quantity 10 g
Synonym Disulfide, bis(phenylmethyl), Benzyl disulfide (8CI), Bis(phenylmethyl) disulfide, 1,4-Diphenyl-2,3-dithiabutane, BDS, Benzyl bisulfide, Dailube S 700, Di(phenylmethyl)disulfide, Dibenzyl disulfide, Dibenzyl disulphide, NSC 677465, NSC 6841, S 700, T 322, α-(Benzyldithio)toluene
InChI Formula InChI=1S/C14H14S2/c1-3-7-13(8-4-1)11-15-16-12-14-9-5-2-6-10-14/h1-10H,11-12H2
SMILES C(SSCc1ccccc1)c2ccccc2
IUPAC Name (benzyldisulfanyl)methylbenzene
Molecular Weight (g/mol) 246.39
Formula Weight 246.0537
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.