missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phosphate, 98%

Product Code. 11382726
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382726 25 g 25g
11362726 1 g 1g
11372726 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382726 Supplier Thermo Scientific Alfa Aesar Supplier No. L08813.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1623-08-1 | C14H15O4P | 278.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1623-08-1
Molecular Formula C14H15O4P
Molecular Weight (g/mol) 278.244
MDL Number MFCD00004775
InChI Key HDFFVHSMHLDSLO-UHFFFAOYSA-N
Synonym dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883
PubChem CID 74189
IUPAC Name dibenzyl hydrogen phosphate
SMILES C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2

Specifications

Melting Point 78°C to 81°C
Quantity 25 g
Beilstein 2055755
Formula Weight 278.24
Percent Purity 98%
Chemical Name or Material Dibenzyl phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.