missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phosphate, 98%

Product Code. 10398643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10398643 1 g Glass bottle 1g
10561652 5 g Glass bottle 5g
10386170 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10398643 Supplier Thermo Scientific Acros Supplier No. 112650010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1623-08-1 | C14H15O4P | 278.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1623-08-1
Molecular Formula C14H15O4P
Molecular Weight (g/mol) 278.23
MDL Number MFCD00004775
InChI Key HDFFVHSMHLDSLO-UHFFFAOYSA-N
Synonym dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883
PubChem CID 74189
IUPAC Name dibenzyl hydrogen phosphate
SMILES C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2

Specifications

Melting Point 76.0°C to 80.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5CH2O)2P(O)OH
Quantity 1 g
Beilstein 06, 439
Formula Weight 278.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dibenzyl phosphate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.