missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl sulfide, 99%, Thermo Scientific™

Product Code. 11496046
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
missing translation for 'unitSize'
100g
25g
This item is not returnable. View return policy

Product Code. 11496046

missing translation for 'mfr': Thermo Scientific Alfa Aesar B22337.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-74-9
Molecular Formula C14H14S
Molecular Weight (g/mol) 214.33
MDL Number MFCD00004784
InChI Key LUFPJJNWMYZRQE-UHFFFAOYSA-N
Synonym dibenzyl sulfide, benzyl sulfide, dibenzyl sulphide, benzyl thioether, benzyl monosulfide, dibenzyl monosulfide, dibenzyl thioether, sulfide, dibenzyl, 1,3-diphenyl-2-thiapropane, benzene, 1,1'-thiobis methylene bis
PubChem CID 10867
IUPAC Name benzylsulfanylmethylbenzene
SMILES C(SCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 47°C to 50°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1911157
Merck Index 14,1145
Formula Weight 214.33
Percent Purity 99%
Chemical Name or Material Dibenzyl sulfide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.