missing translation for 'onlineSavingsMsg'
Learn More

Dibucaine hydrochloride, MedChemExpress

Product Code. 16428528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428528 10 g 10g
16410998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428528 Supplier Medchem Express Supplier No. HYB0552A
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dibucaine hydrochloride (Cinchocaine hydrochloride) is a sodium channel inhibitor. Dibucaine hydrochloride is a potent SChE inhibitor.

Dibucaine hydrochloride (Cinchocaine hydrochloride) is a sodium channel inhibitor. Dibucaine hydrochloride is a potent SChE inhibitor.

Specifications

Chemical Name or Material Dibucaine hydrochloride
CAS 61-12-1
Color White
Purity Grade Notes Research
Molecular Formula C20H30ClN3O2
Quantity 10 g
Synonym Cinchocaine hydrochloride
Solubility Information DMSO : 110 mg/mL (289.53 mM; Need ultrasonic) ∣H2O : 100 mg/mL (263.21 mM; Need ultrasonic)
SMILES O=C(C1=CC(OCCCC)=NC2=CC=CC=C12)NCCN(CC)CC.Cl
Molecular Weight (g/mol) 379.92
Formula Weight 379.92
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.