missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl ether, puriss p.a., ≥99.5% (GC), Honeywell™ Riedel-de-Haën™

Product Code. 15694180
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15694180 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694180 Supplier Honeywell Riedel-de Haën Supplier No. 344621L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., low in aromatic compounds,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 142-96-1
Density 0.764g/mL at 25°C
Flash Point 25°C (77°F)
Ignition Point 365°F
Packaging Glass bottle
Molecular Formula C8H18O
Linear Formula [CH3(CH2)3]2O
MDL Number MFCD00009461
Quantity 1 L
UN Number UN1149
Beilstein 1732752
Synonym di-n-butyl ether, butyl ether, dibutyl ether, butane, 1,1'-oxybis, n-butyl ether, dibutyl oxide, butyl oxide, ether butylique, 1,1'-oxybis butane, di-n-butylether
InChI Key DURPTKYDGMDSBL-UHFFFAOYSA-N
SMILES CCCCOCCCC
IUPAC Name 1-butoxybutane
Molecular Weight (g/mol) 130.23
PubChem CID 8909
Vapor Density 4.48 (vs air)
Formula Weight 130.23g/mol
Vapor Pressure 4.8mmHg (20°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material Dibutyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.