missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phenyl Phosphate, TRC

Product Code. 30382771
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382771 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382771 Supplier Toronto Research Chemicals Supplier No. TRCD429478250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyl Phenyl Phosphate
CAS 2528-36-1
Purity Grade Notes HPLC
Molecular Formula C14 H23 O4 P
Quantity 250 mg
Synonym Butyl phenyl phosphate ((BuO)2(PhO)PO) (6CI, 7CI), Dibutyl phenyl phosphate
InChI Formula InChI=1S/C14H23O4P/c1-3-5-12-16-19(15,17-13-6-4-2)18-14-10-8-7-9-11-14/h7-11H,3-6,12-13H2,1-2H3
SMILES CCCCOP(=O)(OCCCC)Oc1ccccc1
IUPAC Name dibutyl phenyl phosphate
Molecular Weight (g/mol) 286.3
Formula Weight 286.1334
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.