missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phosphate, TRC

Product Code. 30383585
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383585 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383585 Supplier Toronto Research Chemicals Supplier No. TRCD42952025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-n-butylphosphate
CAS 107-66-4
Purity Grade Notes HPLC
Molecular Formula C8 H19 O4 P
Quantity 25 g
Synonym Butyl phosphate, (BuO)2(HO)PO (6CI), DBP, DP 4, DP 4 (coupling agent), Di-n-Butyl phosphate, Dibutyl acid phosphate, Dibutyl hydrogen phosphate, Dibutyl phosphate, HDBP, NSC 203039, NSC 22313
InChI Formula InChI=1S/C8H19O4P/c1-3-5-7-11-13(9,10)12-8-6-4-2/h3-8H2,1-2H3,(H,9,10)
SMILES CCCCOP(=O)(O)OCCCC
IUPAC Name dibutyl hydrogen phosphate
Molecular Weight (g/mol) 210.21
Formula Weight 210.1021
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.