missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl sebacate, 93%

Product Code. 10400831 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10400831

Brand: Thermo Scientific Acros 406650010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 109-43-3 | C18H34O4 | 314.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-43-3
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.46
MDL Number MFCD00027218
InChI Key PYGXAGIECVVIOZ-UHFFFAOYSA-N
Synonym dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs
PubChem CID 7986
IUPAC Name dibutyl decanedioate
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC

Specifications

Melting Point -12°C
Density 0.9400g/mL
Flash Point 167°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3OCO(CH2)8COO(CH2)3CH3
Refractive Index 1.4400 to 1.442
Quantity 1 kg
Specific Gravity 0.94
Formula Weight 314.46
Percent Purity 93%
Physical Form Liquid
Chemical Name or Material Dibutyl sebacate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.