missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Sebacate, TRC

Product Code. 17970065
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970065 50 g 50g
30133003 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17970065 Supplier Toronto Research Chemicals Supplier No. TRCD42952550G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-43-3
Molecular Formula C18 H34 O4
Molecular Weight (g/mol) 314.46
Synonym Decanedioic acid, dibutyl ester (9CI), Sebacic acid, dibutyl ester (6CI,8CI), Bis(n-butyl) sebacate, DBS, Di-n-Butyl sebacate, Dibutyl decanedioate, Dibutyl sebacate, Ergoplast SDB, Kodaflex DBS, NSC 3893, PX 404, Polycizer DBS, Reomol DBS, Sebacic acid di-n-butyl ester, Staflex DBS, Uniflex DBS
IUPAC Name dibutyl decanedioate
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3
Formula Weight 314.2457
Chemical Name or Material Dibutyl Sebacate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.