missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin dilaurate, 94%

Product Code. 11995851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995851 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11995851 Supplier Thermo Scientific Acros Supplier No. 382690050
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 77-58-7 | C32H64O4Sn | 631.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 205.0°C (12.0 mmHg)
Melting Point 16.0°C to 18.0°C
CAS 77-58-7
Color Yellow
Physical Form Oily Liquid
Quantity 5 g
Assay Percent Range 94%
Molecular Formula C32H64O4Sn
Linear Formula [CH3(CH2)3]2Sn[OC(O)CH2(CH2)9CH3]2
Flash Point 149°C
Merck Index 15,3047
Synonym Dodecanoic acid 1, 1';-(dibutylstannylene) ester
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and aceton
Molecular Weight (g/mol) 631.57
Formula Weight 631.57
Percent Purity 94%
Infrared Spectrum Authentic
Packaging Plastic bottle
Refractive Index 1.4690 to 1.4730 (20°C, 589nm)
Specific Gravity 1.05
Density 1.0500g/mL
Chemical Name or Material Dibutyltin dilaurate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dibutyltin dilaurate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H360-May damage fertility or the unborn child.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.