missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin Dilaurate, TRC

Product Code. 30373148
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373148 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373148 Supplier Toronto Research Chemicals Supplier No. TRCD429573250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dibutyltin Dilaurate
CAS 77-58-7
Molecular Formula C32 H64 O4 Sn
Quantity 250 mg
Synonym Dibutyltin dilaurate (6CI), Stannane, dibutylbis(lauroyloxy)- (8CI), Stannane, dibutylbis[(1-oxododecyl)oxy]- (9CI), Tin, dibutylbis(lauroyloxy)- (7CI), Lauric acid, dibutylstannylene deriv. (8CI), A 201, ADK Stab BT 11, Addocat 201, Addocat 210, Advastab T 12P, BL 42A, BNT-CAT 425, BT 11, BT 18, BXX 3778, Baerostab DBTL/C, Butynorate, C 101, C 101 (stabilizer), CAT 11, CAT 11 (catalyst), CM-A, CP, CP (catalyst), Cata-Chek 820, Cotin 200, D 22, D 7103, D 80, D 80 (catalyst), DBTD, DBTDL, DBTL, DBTL-KS 1260, DC 176, DOD-D 82727, DXR 81, DY 12, DY 28, Dabco T 12, Dabco T 12N, Dabco T 12SL, Davainex, Desmorapid Z, Di-n-butyltin dilaurate, Dibutylbis(1-oxododecyloxy)stannane, Dibutylbis(laurato)tin, Dibutylbis(lauroyloxy)tin, Dibutylstannylene dilaurate, Dibutyltin didodecanoate, Dibutyltin n-dodecanoate, Dow Corning 176, Embilizer L 101, Fascat 4202, Foamate S 9, Fomrez SUL 4, Gleck TL, HG 0111, JF-NS-C 010, Jeffcat T 12, K 1 Type A, KS 120, KS 1260, KS 20, Keverkat DBTL 162, Kosmos 19, L 101, LL 101, Lankromark LT 173, Liocat 118, Liocat 119, Mark 1038, Mark BT 1, Mark BT 11, Mark BT 18, Mark DBTL, Meister ZL 4401, Metacure T 12, Metatin 712, Metatin K 712, NSC 2607, Neostann U 100, Niax T 12, Nitto 8501, Octaflow BT 71, Ongrostab BLTM, Reatinor 932, Reaxis C 218, SCAT 1, SCAT 1W, SD-L 101, SM 2014C, SUL 4, Stabilizer D 22, Stanclere DBTL, Stanclere TL, Stann BL, Stann SB 430A, Stavinor 1200SN, T 12, TK 218, TL 10, TL 10 (catalyst), TL 100, TL 1000, TL 1000 (catalyst), TMV 186, TN 12, TN 12 (catalyst), TVS Tin Lau, TVS-TL 700, TVS-TL 800, Tegokat 218, Thermolite T 12, Thorcat 401, Tin dibutyl dilaurate, TinStab BL 277, Tinostat, U 100, U 12, YL 101, YL 101 (catalyst), Yoke T 12, ZT 101, ZT 102, Dodecanoic acid, 1,1'-(dibutylstannylene) ester
InChI Formula InChI=1S/2C12H24O2.2C4H9.Sn/c2*1-2-3-4-5-6-7-8-9-10-11-12(13)14;2*1-3-4-2;/h2*2-11H2,1H3,(H,13,14);2*1,3-4H2,2H3;/q;;;;+2/p-2
SMILES CCCCCCCCCCCC(=O)O[Sn](CCCC)(CCCC)OC(=O)CCCCCCCCCCC
IUPAC Name [dibutyl(dodecanoyloxy)stannyl] dodecanoate
Molecular Weight (g/mol) 631.5582
Formula Weight 632.3827
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.