missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin(IV) Oxide, TRC

Product Code. 30179753
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179753 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179753 Supplier Toronto Research Chemicals Supplier No. TRCD42956010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 818-08-6
Molecular Formula C8 H18 O Sn
Quantity 10 g
Synonym Dibutyltin oxide (6CI), Dibutyloxostannane, Axion CS 2455, Cotin 100, Di-n-butyltin oxide, Di-n-butyltin(II) oxide, Dibutyloxotin, Dibutylstannane oxide, Eurecat 9555, Eurecat 9560, FC 4201, Fascat 4201, Fomrez SUL 11A, Fomrez SUL 11B, Fomrez SUL 11C, MSCAT 01, MSCAT 02, NSC 28130, Neostann U 300, Stann BO, Tegokat 248, Tin dibutyl oxide, U 300, ZT 4201
InChI Formula InChI=1S/2C4H9.O.Sn/c2*1-3-4-2;;/h2*1,3-4H2,2H3;;
SMILES CCCC[Sn](=O)CCCC
IUPAC Name dibutyl(oxo)tin
Molecular Weight (g/mol) 248.94
Formula Weight 250.038
Chemical Name or Material Dibutyltin-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.