missing translation for 'onlineSavingsMsg'
Learn More

Dicarbonylacetylacetonato rhodium(I)

Product Code. 10477402 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10477402

Brand: Thermo Scientific Acros 363955000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14874-82-9
Molecular Formula C7H7O4Rh
Molecular Weight (g/mol) 258.03
MDL Number MFCD00009884
InChI Key BZCAWKOPWNIDOC-FGSKAQBVSA-M
IUPAC Name λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O

Specifications

Melting Point 154.0°C to 156.0°C
Color Green-Red
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: insoluble
Formula Weight 258.04
Physical Form Flakes or Needles
Chemical Name or Material Dicarbonylacetylacetonato rhodium(I)
compliance-icons
Product Identifier
  • Dicarbonylacetylacetonato rhodium(I)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.