missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dichloran Rose-Bengal Chloramphenicol (DRBC) (ISO) Agar Base (Dehydrated)

Product Code. 13265929
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13265929 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13265929 Supplier Thermo Scientific™ Supplier No. CM1148B
  • ONLINE EXCLUSIVE PRICE
    212.00 €
    178.01 € / 500g
    Save 33.99 €
    16% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and enumerate yeast and molds associated with food spoilage with DRBC medium while inhibiting non-target bacteria.

Isolate and enumerate yeasts and molds associated with food spoilage with Thermo Scientific™ Oxoid™ Dichloran Rose-Bengal Chloramphenicol (DRBC) (ISO) Agar Base while adhering to ISO 21527-1: 2008. Dichloran Rose-Bengal Chloramphenicol (DRBC) (ISO) Agar is specially designed to suppress the growth of spreading mold and bacteria while supporting the growth of species that cannot be isolated on Rose Bengal Chloramphenicol Agar or Acidified Potato Dextrose Agar.

Use Dichloran Rose-Bengal Chloramphenicol (DRBC) Agar Base for selective isolation and enumeration of viable yeast and molds in food products with a water activity above 0.95 aW.
  • Adheres to ISO 21527-1: 2008 standards.
  • Selective for viable yeast and moulds in food samples.
DRBC (ISO) is a modification of the original Rose-Bengal Chloramphenicol Medium1 and differs as the Rose-Bengal content has been optimized by adding Dichloran and by changing the pH to 5.6.

The inclusion of chloramphenicol in this medium suppresses bacterial growth, allowing molds, and yeasts to grow. The inhibition of spreading molds and the restriction of their colony size results in improved enumeration and detection. The reduced pH of DRBC (ISO) Agar increases the inhibition of yeasts by the rose-bengal.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Description Dichloran Rose-Bengal Chloramphenicol (DRBC) (ISO) Agar Base (Dehydrated)
Form Powder
Product Type Agar
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.