missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)palladium(II), Pd 36.7%

Product Code. p-7048420
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395988 1 g 1g
11385988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395988 Supplier Thermo Scientific Alfa Aesar Supplier No. 010493.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12107-56-1 | C8H12Cl2Pd | 285.50 g/mol

Dichloro(1,5-cyclooctadiene)palladium(II) is used as catalyst for C-C bond and C-N formation, used in Heck coupling of alkynes with alkenes, Suzuki cross-coupling of aryl bromides , allylic substitution of oximes with allylic acetate and methoxycarbonylation of iodobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12107-56-1
Molecular Formula C8H12Cl2Pd
Molecular Weight (g/mol) 285.50
MDL Number MFCD00012412
InChI Key RRHPTXZOMDSKRS-PHFPKPIQSA-L
Synonym dichloro 1,5-cyclooctadiene palladium ii, cycloocta-1,5-diene dichloropalladium, 1,5-cyclooctadienepalladium ii dichloride, 1z,5z-cycloocta-1,5-diene; palladium 2+ ; dichloride, dichloro 1,5-cyclooctadiene palladium, palladium, dichloro 1,5-cyclooctadiene, dichloro 1,2,5,6-eta-cycloocta-1,5-diene palladium, palladium, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, dichloro 1,5-cyclooctadiene palladium ii ,, dichloro 1,5-cyclooctadiene palladium ii 1g
PubChem CID 6436380
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;palladium(2+);dichloride
SMILES [Cl-].[Cl-].[Pd++].C1C\C=C/CC\C=C/1

Specifications

Color Yellow
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in dichloromethane
Formula Weight 285.49
Concentration or Composition (by Analyte or Components) Pd 36.7%
Physical Form Crystalline
Chemical Name or Material Dichloro(1,5-cyclooctadiene)palladium(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.