missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)platinum(II), Pt 51.6-52.6%

Product Code. 11387527
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387527 0.25 g 0.25g
11397527 1 g 1g
11377527 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387527 Supplier Thermo Scientific Alfa Aesar Supplier No. 012739.01
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12080-32-9 | C8H12Cl2Pt | 374.17 g/mol

it is used in Hydrosilyation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12080-32-9
Molecular Formula C8H12Cl2Pt
Molecular Weight (g/mol) 374.17
MDL Number MFCD00012413
InChI Key VVAOPCKKNIUEEU-PHFPKPIQSA-L
Synonym dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
PubChem CID 6436378
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
SMILES Cl[Pt]Cl.C1C\C=C/CC\C=C/1

Specifications

Melting Point 250°C (decomposition)
Quantity 0.25 g
Sensitivity Hygroscopic
Solubility Information Soluble in dichloromethane
Formula Weight 374.18
Concentration or Composition (by Analyte or Components) Pt 51.6 to 52.6%
Physical Form Solid
Chemical Name or Material Dichloro(1,5-cyclooctadiene)platinum(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.