missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II)

Product Code. 11307238
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11307238

Brand: Thermo Scientific Alfa Aesar 030167.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15629-92-2 | C27H26Cl2NiP2 | 542.04 g/mol

Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II) is used as an effective catalyst for Kumada coupling and Suzuki reactions. It also catalyzes other reactions to convert enol ethers, dithioacetals and vinyl sulfides to olefins. Further, it is used as a catalyst for preparation of block-copolythiophenes and solid state metathesis polycondensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15629-92-2
Molecular Formula C27H26Cl2NiP2
Molecular Weight (g/mol) 542.04
MDL Number MFCD00015318
InChI Key REQVOKPBMSCTNI-UHFFFAOYSA-L
Synonym 1,3-Bis(diphenylphosphino)propane nickel(II) chloride
PubChem CID 131675641
IUPAC Name 3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
SMILES Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213°C (decomposition)
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 542.03
Physical Form Powder
Chemical Name or Material Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.