missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(p-cymene)ruthenium(II) dimer, 98%

Product Code. 11399665
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Unit Size:
10g
2g
500mg
Product Code. Quantity unitSize
11399665 500 mg 500mg
11309675 2 g 2g
11319675 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 11399665

Brand: Thermo Scientific Alfa Aesar L19126.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52462-29-0 | C20H28Cl4Ru2 | 612.384 g/mol

Dichloro(p-cymene)ruthenium(II) dimer is used as hydrosilylation catalyst and also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloro(p-cymene)ruthenium(II) dimer is used as hydrosilylation catalyst and also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52462-29-0
Molecular Formula C20H28Cl4Ru2
Molecular Weight (g/mol) 612.384
MDL Number MFCD00064793
InChI Key LAXRNWSASWOFOT-UHFFFAOYSA-J
Synonym dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer
PubChem CID 10908223
IUPAC Name dichlororuthenium;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl

Specifications

Quantity 500 mg
Solubility Information Insoluble in water.
Formula Weight 612.4
Percent Purity 98%
Chemical Name or Material Dichloro(p-cymene)ruthenium(II) dimer
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.