missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, TRC

Product Code. 30387387
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387387 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387387 Supplier Toronto Research Chemicals Supplier No. TRCD438770250MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer
CAS 12354-84-6
Molecular Formula C20H30Cl4Ir2
Quantity 250 mg
Synonym (Pentamethylcyclopentadienyl)iridium dichloride dimer
InChI Formula InChI=1S/2C10H15.4ClH.2Ir/c2*1-6-7(2)9(4)10(5)8(6)3;;;;;;/h2*1-5H3;4*1H;;/q2*-1;;;;;2*+3/p-4
SMILES [Cl-][Ir+3]12345([Cl-][Ir+3]6789([Cl-])([Cl-]1)C=%10(C)C9(C)=C8(C)[C-]7(C)C%106C)C=%11(C)C5(C)=C4(C)[C-]3(C)C%112C
Molecular Weight (g/mol) 796.7
Formula Weight 796.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.