missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic acid, technical, Thermo Scientific™

Product Code. 17525625 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17525625 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17525625 Supplier Thermo Scientific Acros Supplier No. 473140025
  • ONLINE EXCLUSIVE PRICE
    248.00 €
    229.00 € / 2.5 L
    Save 19.00 €
    7% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector DICHLOROACETICACID
CAS 79-43-6
Color Clear colorless to dark yellow
Flash Point >112°C
Infrared Spectrum Conforms
Assay Percent Range 98.5 to 101.5% (Titration with NaOH)
Packaging Glass Bottle
Molecular Formula C2H2Cl2O2
Linear Formula Cl2CHCO2H
Refractive Index 1.4650 to 1.4670 (20°C, 589 nm)
MDL Number 00004223
Quantity 2.5 L
Solubility Information Soluble in water; Miscible with alcohol, ether and ethanol; Soluble in acetone
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
SMILES OC(=O)C(Cl)Cl
IUPAC Name 2,2-dichloroacetic acid
Molecular Weight (g/mol) 128.94
Physical Form Liquid
Chemical Name or Material Dichloroacetic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichloroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity on or via lactation
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H350-May cause cancer.
  • H360F-May damage fertility.
  • H362-May cause harm to breast-fed children.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Dichloroacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.