missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dichlorobis(pyridine)palladium(II), 98%

Product Code. 11459117
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11459117 Supplier Alfa Aesar™ Supplier No. B20663.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14872-20-9
Molecular Formula C10H10Cl2N2Pd
MDL Number MFCD00792248
Synonym Bis(pyridine)palladium(II) chloride

Specifications

Quantity 1 g
Formula Weight 335.53
Percent Purity 98%
Chemical Name or Material Dichlorobis(pyridine)palladium(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.