missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylgermane, 95+%

Product Code. 10195470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10195470 1 g Glass bottle 1g
10311104 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10195470 Proveedor Thermo Scientific Acros N.º de proveedor 316500010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1529-48-2 | C2H6Cl2Ge | 173.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1529-48-2
Molecular Formula C2H6Cl2Ge
Molecular Weight (g/mol) 173.57
MDL Number MFCD00013588
InChI Key YQECBLVSMFAWIZ-UHFFFAOYSA-N
Synonym dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
PubChem CID 73724
IUPAC Name dichloro(dimethyl)germane
SMILES C[Ge](C)(Cl)Cl

Especificaciones

Melting Point -22°C
Color Colorless
Density 1.5000g/mL
Boiling Point 123°C
Flash Point 32°C
Assay Percent Range 95% min
Packaging Glass bottle
Linear Formula (CH3)2GeCl2
Refractive Index 1.4590 to 1.461
Quantity 1 g
UN Number 2920
Specific Gravity 1.5
Solubility Information Solubility in water: reacts
Formula Weight 173.57
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material Dichlorodimethylgermane
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.