missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylgermane, 95+%

Product Code. 10311104 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311104 5 g Glass bottle 5g
10195470 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10311104 Supplier Thermo Scientific Acros Supplier No. 316500050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1529-48-2 | C2H6Cl2Ge | 173.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1529-48-2
Molecular Formula C2H6Cl2Ge
Molecular Weight (g/mol) 173.57
MDL Number MFCD00013588
InChI Key YQECBLVSMFAWIZ-UHFFFAOYSA-N
Synonym dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
PubChem CID 73724
IUPAC Name dichloro(dimethyl)germane
SMILES C[Ge](C)(Cl)Cl

Specifications

Melting Point -22°C
Color Colorless
Density 1.5000g/mL
Boiling Point 123°C
Flash Point 32°C
Assay Percent Range 95% min
Packaging Glass bottle
Linear Formula (CH3)2GeCl2
Refractive Index 1.4590 to 1.461
Quantity 5 g
UN Number 2920
Specific Gravity 1.5
Solubility Information Solubility in water: reacts
Formula Weight 173.57
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material Dichlorodimethylgermane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.