Learn More
Dichlorodimethylsilane, 98+%
Description
Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with alcohols, amines and strong bases.
Spécification
Spécification
| Boiling Point | 70°C to 71°C |
| Melting Point | ∼-76°C |
| Quantity | 100 g |
| Linear Formula | (CH3)2SiCl2 |
| UN Number | UN1162 |
| Beilstein | 605287 |
| Flash Point | −5°C (23°F) |
| Solubility Information | Immiscible with water. |
| Formula Weight | 129.06 |
| Percent Purity | 99% |
| Afficher plus |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.