missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodimethylsilane, 99+%, AcroSeal™

Product Code. 10217647 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10217647 100 mL AcroSeal™ Glass Bottle 100mL
10777708 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217647 Supplier Thermo Scientific Acros Supplier No. 430881000
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-78-5
Molecular Formula C2H6Cl2Si
Molecular Weight (g/mol) 129.06
MDL Number MFCD00000491
InChI Key LIKFHECYJZWXFJ-UHFFFAOYSA-N
Synonym dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
PubChem CID 6398
IUPAC Name dichloro(dimethyl)silane
SMILES C[Si](C)(Cl)Cl

Specifications

Melting Point -76.0°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 70.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)2SiCl2
Refractive Index 1.4030 to 1.4050
Quantity 100 mL
Specific Gravity 1.06
Solubility Information Solubility in water: reacts. Other solubilities: soluble in proton-free solvents (care must be,taken with ketones and esters),soluble in benzene and ether,decomposes in ethanol
Viscosity 0.6 mPa.s (25°C)
Formula Weight 129.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dichlorodimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorodimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.