missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodipropyltin, TRC

Product Code. 17954655
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954655 1 g 1g
17982695 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17954655 Supplier Toronto Research Chemicals Supplier No. TRCD4342701G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 867-36-7
Molecular Formula C6 H14 Cl2 Sn
Molecular Weight (g/mol) 275.79
Synonym Dichlorodipropyltin, Di-n-propyltin-dichloride
IUPAC Name dichloro(dipropyl)stannane
SMILES CCC[Sn](Cl)(Cl)CCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C3H7.2ClH.Sn/c2*1-3-2;;;/h2*1,3H2,2H3;2*1H;/q;;;;+2/p-2
Formula Weight 275.9495
Percent Purity >95%
Chemical Name or Material Di-n-propyl-tin-dichloride
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.