missing translation for 'onlineSavingsMsg'
Learn More

Dichloroisocyanuric acid sodium salt, 98%

Product Code. 10451631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10451631 500 g 500g
10712251 25 g 25g
15274674 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10451631 Supplier Thermo Scientific Acros Supplier No. 204115000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger | CAS: 2893-78-9 | C3HCl2N3NaO3 | 220.949 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2893-78-9
Molecular Formula C3HCl2N3NaO3
Molecular Weight (g/mol) 220.949
MDL Number MFCD00006036
InChI Key UNWRHVZXVVTASG-UHFFFAOYSA-N
Synonym sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
PubChem CID 86657659
IUPAC Name 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
SMILES C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Specifications

Color White
pH 6 to 7 (1% aq. soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Formula Weight 219.95
Percent Purity ≥97.5% (on dry substance)
Physical Form Crystals or Granules
Chemical Name or Material Dichloroisocyanuric acid sodium salt, 98%
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichloroisocyanuric acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Mention de danger
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.