missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99+%, Extra Pure, Stabilised with Amylene, SLR, Fisher Chemical™

Product Code. 10580141 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal can
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10580141 500 mL Amber Glass Bottle 500mL
10127611 1 L Amber Glass Bottle 1 L
10458210 2.5 L Amber Glass Bottle 2.5 L
10774941 2.5 L Plastic-coated Glass Bottle 2.5 L
10550641 5 L HDPE Bottle 5 L
10273850 5 L Metal can 5 L
10590141 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10580141 Supplier Fisher Chemical Supplier No. D/1850/08
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 350mbar at 20°C
Viscosity 0.43 mPaS at 20°C
Boiling Point 39°C
Formula Weight 84.93g/mol
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.