missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15677830
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
10 L
2.5 L
20 L
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15677830 1 L Glass Bottle 1 L
15687830 2.5 L Glass Bottle 2.5 L
15607840 6 x 1 L Glass Bottle Pack of 6
15667830 10 L Stainless Steel Can 10 L
15697830 20 L Stainless Steel Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15677830 Supplier Honeywell-Riedel-de Haen Supplier No. D651001L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, contains 40-150 amylene as stabilizer,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C (literature)
CAS Min % ≥99.0000%
CAS Max % ≥99.0000%
Density 1.325 g/mL at 25 °C(lit.)
Quantity 1 L
Packaging Glass Bottle
Linear Formula CH2Cl2
UN Number UN1593
Water max. 0.02%
Beilstein 1730800
Boiling Point 39°C to 41°C
Formula Weight 84.93
Percent Purity >99.5%
Grade ACS
Chemical Name or Material Dichloromethane, ACS Reagent, contains 40-150 amylene as stabilizer, ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.