missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riedel-de Haën Dichloromethane CHROMASOLV™
Description
- Low impurity gradient LC baseline.
Specifications
Specifications
| pH | Not applicable |
| Melting Point | -97°C |
| CAS | 75-09-2 |
| CAS Min % | ≥99.00% |
| Color | Colorless |
| Density | 1.32 to 1.33 g/cm3 (20°C) |
| Boiling Point | 39°C to 41°C (1.013 hPa) |
| Quantity | 45 L |
| Flash Point | Not applicable |
| Packaging | Returnable Stainless Steel Drum |
| Show More |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?