missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, CHROMASOLV™, for analysis of dioxins, furans and PCB, Solstice

Product Code. 15684170
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15684170 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684170 Supplier Honeywell-Chromasolv Supplier No. 344112.5L
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CHROMASOLV™, for analysis of dioxins, furans and PCB

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-09-2
Density 1.325g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.22
Ignition Point 1223°F
Molecular Formula CH2Cl2
Vapor Pressure 24.45 psi (55°C), 6.83 psi (20°C)
MDL Number MFCD00000881
UN Number UN1593
Beilstein 1730800
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
PubChem CID 6344
ChEBI CHEBI:15767
Formula Weight 84.93g/mol
Percent Purity ≥99.8% (GC)
Grade CHROMASOLV
Chemical Name or Material Dichloromethane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.