missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane CHROMASOLV™, for HPLC, ≥99.8%, contains amylene as stabilizer, Solstice

Product Code. 15684540
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
100mL
2.5 L
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15684540 2.5 L 2.5 L
15664540 100 mL 100mL
15674540 1 L 1 L
15614550 6 x 1 L Pack of 6
15694540 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15684540 Supplier Honeywell-Chromasolv Supplier No. 348562.5L
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Product filtered through a 0.2μm filter

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Density 1.325g/mL at 25°C
Quantity 2.5 L
Refractive Index n20/D 1.424
Linear Formula CH2Cl2
Vapor Pressure 6.83 psi (20°C)
Beilstein 1730800
Boiling Point 39.8°C to 40°C
Formula Weight 84.93g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material Dichloromethane, for HPLC, contains amylene as stabilizer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methylene chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.