missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d{2}, 99.5% (Isotopic)

Product Code. 15413177
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413177 25 g 25g
15403177 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15413177 Supplier Thermo Scientific Alfa Aesar Supplier No. A18036.14
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1665-00-5 | CH2Cl2 | 86.94 g/mol

NMR solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

NMR solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.94
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
IUPAC Name dichloro(²H₂)methane
SMILES [2H]C([2H])(Cl)Cl

Specifications

Quantity 25 g
Boiling Point 39°C to 41°C
Percent Purity 99.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.