missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d{2}, 99.9% (Isotopic)

Product Code. 11384418
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384418 2 Ea. Pack of 2
11374418 10 Ea. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11384418 Supplier Thermo Scientific Alfa Aesar Supplier No. 042335.SA
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Acts as a solvent in Nuclear magnetic resonance (NMR) spectroscopy. | CAS: 1665-00-5

Dichloromethane-d{2} acts as a solvent in Nuclear magnetic resonance (NMR) spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
MDL Number MFCD00000882
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d

Specifications

Density 1.35
Packaging Prescored ampoules, 0.75mL/ampoule
Quantity 2 Ea.
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol,ether and dimethyl formamide.
Formula Weight 86.95
Percent Purity 99.9%
Physical Form Liquid
Chemical Name or Material Dichloromethane-d2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.