missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane-d2, for NMR, packaged in 0.50 ml ampoules, 100.0 atom % D, ACROS Organics™

Product Code. 10378490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 x 0.50 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10378490 10 x 0.50 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10378490 Supplier Thermo Scientific Acros Supplier No. 217430050
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Density 1.3600g/mL
Quantity 10 x 0.50 mL
CAS 1665-00-5
Molecular Formula CH2Cl2
Linear Formula CD2Cl2
MDL Number MFCD00000882
Beilstein 01, IV, 39
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
SMILES C(Cl)Cl
IUPAC Name dichloro(dideuterio)methane
Molecular Weight (g/mol) 86.939
PubChem CID 160586
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2, 100.0 atom % D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.