missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylborane, 97%

Product Code. 10116000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10116000 5 g Glass bottle 5g
10083953 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10116000 Supplier Thermo Scientific Acros Supplier No. 318550050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 873-51-8 | C6H5BCl2 | 158.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-51-8
Molecular Formula C6H5BCl2
Molecular Weight (g/mol) 158.82
MDL Number MFCD00013612
InChI Key NCQDQONETMHUMY-UHFFFAOYSA-N
Synonym phenylboron dichloride, borane, dichlorophenyl, dichloro phenyl borane, phenyldichloroborane, phenylboron dibromide, dichlorophenylboron, dichloro-phenyl borane, dichlorophenylborane, bis chloranyl-phenyl-borane, dichloro-phenyl-boron
PubChem CID 136678
IUPAC Name dichloro(phenyl)borane
SMILES B(C1=CC=CC=C1)(Cl)Cl

Specifications

Melting Point 7.0°C
Color Colorless
Density 1.2200g/mL
Boiling Point 175.0°C
Flash Point −14°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5BCl2
Refractive Index 1.5450 to 1.5470
Quantity 5 g
Specific Gravity 1.22
Formula Weight 158.82
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichlorophenylborane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichlorophenylborane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Dichlorophenylborane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.