missing translation for 'onlineSavingsMsg'
Learn More

Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

Product Code. 11352909
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352909 100 mg 100mg
11362909 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352909 Supplier Thermo Scientific Alfa Aesar Supplier No. H27752.MC
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 92220-65-0 | C22H16ClIrN2 | 536.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92220-65-0
Molecular Formula C22H16ClIrN2
Molecular Weight (g/mol) 536.05
MDL Number MFCD07784515,MFCD28144567
InChI Key ORDROLZETYMBQO-UHFFFAOYSA-M
Synonym bis 2-pyridin-2-yl phenyl iridium iii chloride
PubChem CID 132274317
IUPAC Name iridium(3+);2-phenylpyridine;chloride
SMILES [Cl-].[Ir+3].C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1

Specifications

Quantity 100 mg
Formula Weight 1072.09
Chemical Name or Material Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.