missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclofenac-13C6 Sodium Salt (may contain up to 3% unlabelled), TRC
Specifications
Specifications
| Chemical Name or Material | Diclofenac-13C6 Sodium Salt |
| Molecular Formula | 13C6 C8 H10 Cl2 N Na O2 |
| Quantity | 5 mg |
| Synonym | 2-((2,6-Dichlorophenyl)aminobenzeneacetic Acid-13C6 Sodium Salt, [o-(2,6-Dichloroanilino)phenyl]acetic Acid Monosodium Salt-13C6, Sodium Diclofenac-13C6, Sorelmon-13C6, Tsudohmin-13C6, Valetan-13C6, Voldal-13C6, Voltaren-13C6, Voltaren Ophtha-13C6, Benfofen-13C6, Delphimix-13C6, Diacron-13C6, Naclof-13C6, Naclofen-13C6, Neriodin-13C6, Novapirina-13C6, Orthofen-13C6, Orthophen-13C6 |
| InChI Formula | InChI=1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1/i1+1,2+1,4+1,7+1,9+1,12+1; |
| SMILES | [Na]OC(=O)C[13c]1[13cH][13cH][13cH][13cH][13c]1Nc2c(Cl)cccc2Cl |
| IUPAC Name | [2-[2-(2,6-dichloroanilino)(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl]acetyl]oxysodium |
| Molecular Weight (g/mol) | 324.086 |
| Formula Weight | 323.019 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?