missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Sodium Salt (1 mg/ml in Methanol), TRC

Product Code. 30405548
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405548 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405548 Supplier Toronto Research Chemicals Supplier No. TRCKIT34661X1ML
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diclofenac sodium
CAS 15307-79-6
Molecular Formula C14 H10 Cl2 N O2 . Na
Quantity 1x1 mL
Synonym Acetic acid, [o-(2,6-dichloroanilino)phenyl]-, monosodium salt (8CI), Benzeneacetic acid, 2-[(2,6-dichlorophenyl)amino]-, monosodium salt (9CI), 2-(2,6-Dichloroanilino)phenylacetic acid sodium salt, 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetic acid sodium salt, Abitren, Allvoran, Assaren, Benfofen, Bolabomin, Daispas, Declophen, Dedolor, Deflamat, Delphimix, Diacron, Dichronic, Diclo-Phlogont, Diclo-Puren, Diclobene, Diclobenin, Dicloberl, Dicloberl Retard, Diclodyn, Diclofen SR 100, Diclofenac retard, Diclofenac sodium, Diclofenac sodium salt, Diclofenac-Na Emulgel, Diclofenacsodium Emulgel, Dicloflex, Diclogesic, Diclokalium, Diclomol, Diclon, Diclophenac sodium, Dicloran CP, Dicloran Plus, Diclord, Diclorep, Dicloreum, Difene, Dikloron, Diklovit, Dolaut, Dolobasan, Duravolten, Dyclo, Dyloject, Dynapar, Effekton, Evofenac, Feloran, Fortfen, GP 45840, Hyanalgese D, Inflaban, Kriplex, Modifenac, N-(2,6-Dichlorophenyl)-o-aminophenylacetic acid sodium salt, Naclof, Naclofen, Naclofen Duo, Neriodin, Neurodiclovit, Neurofenac, Novapirina, Olfen, Orthofen, Orthophen, Panamor, Phlogin, Primofenac, Profenac, Prophenatin, Refen, Refen retard, Rhumalgan, SR 318B, Saffrac, Sodium [o-(2,6-dichloroanilino)phenyl]acetate, Sodium diclofenac, Sorelmon, Tsudohmin, Valetan, Voldal, Voltaren, Voltaren Ophtha, Voltaren Ophtha CD, Voltarol, Voltral, Voveran, Voveran Plus, Xenid, [o-(2,6-Dichloroanilino)phenyl]acetic acid sodium salt, diclotard
InChI Formula InChI=1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1
SMILES [Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]acetate
Molecular Weight (g/mol) 318.13
Formula Weight 316.9986
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.