Learn More
Dicloxacillin sodium salt monohydrate, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J61581.03
| Quantity | 1g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 13412-64-1 | |
| 513.342 | |
| ASMFXKXNAWVTAY-MBGFYOAHSA-N | |
| (2S,5R,6R)-6-[[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate |
| C19H21Cl2N3NaO6S | |
| MFCD00056865 | |
| 131673926 | |
| [HH].CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] |
Specifications
| 13412-64-1 | |
| MFCD00056865 | |
| 14,3084 | |
| ASMFXKXNAWVTAY-MBGFYOAHSA-N | |
| (2S,5R,6R)-6-[[3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate | |
| 131673926 | |
| Dicloxacillin sodium salt |
| C19H21Cl2N3NaO6S | |
| 1g | |
| Soluble in water | |
| [HH].CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] | |
| 513.342 | |
| 492.33 |
Safety and Handling
- Dicloxacillin sodium salt monohydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.