missing translation for 'onlineSavingsMsg'
Learn More

Dicyandiamide, 99.5%

Product Code. 10639833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
1 kg
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
2.5kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10639833 25 g Plastic bottle 25g
10242462 5 g Glass Bottle 5g
10284040 1 kg Plastic bottle 1kg
10182470 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10639833 Supplier Thermo Scientific Acros Supplier No. 113810250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 461-58-5 | C2H4N4 | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
SMILES NC(N)=NC#N

Specifications

Melting Point 208.0°C to 211.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Packaging Plastic bottle
Quantity 25 g
Fieser 01,229
Merck Index 15, 3103
Solubility Information Solubility in water: 32g/L (20°C). Other solubilities: 38g/L in methanol (20°C)
IUPAC Name N''-cyanoguanidine
Formula Weight 84.08
Percent Purity 99.5%
Physical Form Crystalline Powder
Chemical Name or Material Dicyandiamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.