missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine, ≥99.5% (GC), Honeywell Fluka™

Product Code. 15635130 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
500mL
Product Code. Quantity Packaging unitSize
15635130 100 mL Glass Bottle 100mL
15645130 500 mL Glass Bottle 500mL
2 options
This item is not returnable. View return policy

Product Code. 15635130

Brand: Honeywell-Fluka 36620100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for titrimetric determination of isocyanates,≥99.5% (GC)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-83-7
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.32
MDL Number MFCD00011658
InChI Key XBPCUCUWBYBCDP-UHFFFAOYSA-N
Synonym dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
PubChem CID 7582
ChEBI CHEBI:34694
IUPAC Name N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1

Specifications

Melting Point -2°C
Color Colorless to Yellow
Density 0.912g/mL at 20°C
Boiling Point 117°C to 120°C
Flash Point 96°C (204.8°F)
Packaging Glass Bottle
Linear Formula (C6H11)2NH
Refractive Index n20/D 1.484
Quantity 100 mL
UN Number UN2565
Beilstein 605923
Vapor Density 6 (vs air)
Formula Weight 181.32g/mol
Vapor Pressure 12mmHg (37.7°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material Dicyclohexylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.