missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine, ≥99.5% (GC), Honeywell Fluka™

Product Code. 15645130 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15645130 500 mL Glass Bottle 500mL
15635130 100 mL Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15645130 Supplier Honeywell-Fluka Supplier No. 36620500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for titrimetric determination of isocyanates,≥99.5% (GC)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-83-7
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.32
MDL Number MFCD00011658
InChI Key XBPCUCUWBYBCDP-UHFFFAOYSA-N
Synonym dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
PubChem CID 7582
ChEBI CHEBI:34694
IUPAC Name N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1

Specifications

Melting Point -2°C
Color Colorless to Yellow
Density 0.912g/mL at 20°C
Boiling Point 117°C to 120°C
Flash Point 96°C (204.8°F)
Packaging Glass Bottle
Linear Formula (C6H11)2NH
Refractive Index n20/D 1.484
Quantity 500 mL
UN Number UN2565
Beilstein 605923
Vapor Density 6 (vs air)
Formula Weight 181.32g/mol
Vapor Pressure 12mmHg (37.7°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material Dicyclohexylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.