missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylamine, 99+%

Product Code. 10174390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174390 5 mL Glass bottle 5mL
10532174 100 mL Glass bottle 100mL
10721271 500 mL Glass bottle 500mL
10636012 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10174390 Supplier Thermo Scientific Acros Supplier No. 181050050
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101-83-7 | C12H23N | 181.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-83-7
Molecular Formula C12H23N
Molecular Weight (g/mol) 181.32
MDL Number MFCD00011658
InChI Key XBPCUCUWBYBCDP-UHFFFAOYSA-N
Synonym dicyclohexylamine, dcha, dicha, cyclohexanamine, n-cyclohexyl, n,n-dicyclohexylamine, dodecahydrodiphenylamine, dicyklohexylamin, n,n-diclohexylamine, n-cyclohexyl-cyclohexylamine, dicyklohexylamin czech
PubChem CID 7582
ChEBI CHEBI:34694
IUPAC Name N-cyclohexylcyclohexanamine
SMILES C1CCC(CC1)NC1CCCCC1

Specifications

Melting Point -2.0°C
Color White
Density 0.9100g/mL
Boiling Point 256.0°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C6H11)2NH
Refractive Index 1.4832 to 1.4852
Quantity 5 mL
Beilstein 12,6
Fieser 01,231
Merck Index 15,3106
Specific Gravity 0.91
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 7.4 mPa.s (20°C)
Formula Weight 181.32
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Dicyclohexylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dicyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.